Skip to Content
Merck
All Photos(1)

Key Documents

PZ0386

Sigma-Aldrich

PF-06456384 trihydrochloride

≥98% (HPLC)

Synonym(s):

3-Cyano-4-[[3-[2-[[[2-(4-piperidinyl)ethyl]amino]methyl]-4-pyridinyl]-3′-(trifluoromethyl)[1,1′-biphenyl]-4-yl]oxy]-N-1,2,4-thiadiazol-5-yl-benzenesulfonamide trihydrochloride, Benzenesulfonamide, 3-cyano-4-[[3-[2-[[[2-(4-piperidinyl)ethyl]amino]methyl]-4-pyridinyl]-3′-(trifluoromethyl)[1,1′-biphenyl]-4-yl]oxy]-N-1,2,4-thiadiazol-5-yl-, hydrochloride (1:3)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H32F3N7O3S2 · 3HCl
CAS Number:
Molecular Weight:
829.18
UNSPSC Code:
12352202
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

room temp

SMILES string

O=S(C(C=C1)=CC(C#N)=C1OC2=C(C3=CC(CNCCC4CCNCC4)=NC=C3)C=C(C5=CC=CC(C(F)(F)F)=C5)C=C2)(NC6=NC=NS6)=O

InChI key

WSPHMGORSRQULF-UHFFFAOYSA-N

Biochem/physiol Actions

PF-06456384 is a highly potent and selective inhibitor against voltage-gated sodium channel NaV1.7 (SCN9A; IC50 = 0.01, <0.1, 75 nM against human, mouse, rat Nav1.7, respectively, by conventional patch clamp; human Nav1.7 IC50 = 0.58 nM by PatchExpress electrophysiology) with >300-fold selectivity over other human NaVs (IC50 = 3 nM/NaV1.2, 5.8 nM/NaV1.6, 75 nM/NaV1.7, 314 nM/NaV1.1, 1.45 μM/NaV1.4, 2.59 μM/NaV1.5, 6.44 μM/NaV1.3, 26 μM/NaV1.8).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R Ian Storer et al.
Bioorganic & medicinal chemistry letters, 27(21), 4805-4811 (2017-10-17)
The discovery and selection of a highly potent and selective Na

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service