Key Documents
CLS430597
Corning® non-treated culture dishes
D × H 150 mm × 25 mm
Synonym(s):
cell culture dishes, cell culture plates, culture dish
About This Item
UNSPSC Code:
41121812
NACRES:
NB.13
material
polystyrene
round
sterility
sterile
packaging
sleeve of 5
manufacturer/tradename
Corning 430597
D × H
150 mm × 25 mm
surface area
148 cm2
General description
  • Manufactured from optically-clear virgin polystyrene
  • Not treated for cell attachment, for applications where cell attachment is not desired
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
The 150 and 245 mm culture dishes make excellent carriers and incubator trays for 35 and 60 mm dishes. This avoids spills and lowers the risk of contamination.
Legal Information
Corning is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)