Skip to Content
Merck
All Photos(1)

Key Documents

S-059

Supelco

AM2201 4-Hydroxypentyl metabolite solution

100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H22FNO2
CAS Number:
Molecular Weight:
375.44
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

form

liquid

feature

(Snap-N-Spike®)

packaging

ampule of 1 mL

manufacturer/tradename

Cerilliant®

drug control

Narcotic Licence Schedule E (Switzerland)

concentration

100 μg/mL in methanol

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

application(s)

forensics and toxicology
forensics and toxicology

format

single component solution

storage temp.

−20°C

SMILES string

O=C(C1=CN(CCCC(CF)O)C2=C1C=CC=C2)C3=CC=CC4=C3C=CC=C4

InChI

1S/C24H22FNO2/c25-15-18(27)9-6-14-26-16-22(20-11-3-4-13-23(20)26)24(28)21-12-5-8-17-7-1-2-10-19(17)21/h1-5,7-8,10-13,16,18,27H,6,9,14-15H2

InChI key

HTFNNXALSQKDIS-UHFFFAOYSA-N

General description

A primary urinary metabolite of AM2201, a synthetic cannabinoid found in blends of the herbal mixture Spice. This Certified Spiking Solution® is suitable as starting material for calibrators and controls in GC/MS and LC/MS Spice cannabinoid testing methods for forensic analysis, clinical toxicology, and urine drug testing applications.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
CERTIFIED SPIKING SOLUTION is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service