Skip to Content
Merck
All Photos(1)

Key Documents

860055P

Avanti

Ganglioside GD1a

Avanti Research - A Croda Brand 860055P, powder

Synonym(s):

Disialoganglioside-GD1a (porcine brain, diammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C84H154N6O39
CAS Number:
Molecular Weight:
1872.14
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

Assay

≥99% (TLC)

form

powder

packaging

pkg of 1 × 1 mg (860055P-1MG)
pkg of 1 × 5 mg (860055P-5MG)

manufacturer/tradename

Avanti Research - A Croda Brand 860055P

lipid type

lipid extracts
sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](/C=C/CCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCCCCCCCC)=O)CO[C@H]1[C@H](O)C(O)[C@H](O[C@@H]2O[C@H](CO)[C@@H]([C@H](O[C@@]3(C([O-])=O)O[C@@H]([C@H](O)[C@H](O)CO)[C@@](NC(C)=O)([H])[C@@H](O)C3)[C@H]2O)O[C@@H]4O[C@@H]([C@@H]([C@H](O[C@@H]5O[C@H](CO)[C

InChI

1S/C86H152N4O39.2Na/c1-6-8-10-12-14-16-18-20-22-24-26-28-30-32-34-36-51(100)50(90-60(105)37-35-33-31-29-27-25-23-21-19-17-15-13-11-9-7-2)46-118-80-69(111)67(109)72(57(43-94)120-80)123-82-71(113)78(129-86(84(116)117)39-53(102)62(88-48(4)98)77(128-86)65(107)55(104)41-92)74(59(45-96)122-82)124-79-63(89-49(5)99)75(66(108)56(42-93)119-79)125-81-70(112)68(110)73(58(44-95)121-81)126-85(83(114)115)38-52(101)61(87-47(3)97)76(127-85)64(106)54(103)40-91;;/h34,36,50-59,61-82,91-96,100-104,106-113H,6-33,35,37-46H2,1-5H3,(H,87,97)(H,88,98)(H,89,99)(H,90,105)(H,114,115)(H,116,117);;/q;2*+1/p-2/b36-34+;;/t50-,51+,52?,53?,54-,55-,56?,57?,58?,59?,61+,62+,63?,64-,65-,66?,67?,68?,69?,70?,71?,72?,73?,74?,75?,76?,77?,78?,79?,80?,81?,82?,85?,86?;;/m0../s1

InChI key

CWLDKTAXQZEVQN-CUEXFKMESA-L

General description

Ganglioside GD1a is an anionic brain ganglioside. It is localized in the inner and outer membrane of nuclear envelope of the neuronal cells. GD1a contains a neutral tetrasaccharide core, with one or two sialic acid on the internal galactose. It also has an additional sialic acid on the non-reducing terminal of galactose.

Packaging

5 mL Amber Glass Screw Cap Vial (860055P-1MG)
5 mL Amber Glass Screw Cap Vial (860055P-5MG)

Other Notes

Structure, molecular formula, and molecular weight are for a representative form of the material.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ganglioside GD1a increases the excitability of voltage-dependent sodium channels
Salazar B C, et al.
Brain Research, 1021(2), 151-158 (2004)
Sialidase occurs in both membranes of the nuclear envelope and hydrolyzes endogenous GD1a
Wang J, et al.
Journal of Neurochemistry, 111(2), 547-554 (2009)
Biosynthesis of the major brain gangliosides GD1a and GT1b
Sturgill E R, et al.
Glycobiology, 22(10), 1289-1301 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service