Skip to Content
Merck
All Photos(1)

Key Documents

WHA1810090

Whatman® borosilicate glass TCLP testing filters 0.7 μm pore size

white, acid-treated, low metal, 50 ea, 90 mm diam

Synonym(s):

Whatman filtration, Z740780, metal filtration, tclp circles

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
40161507
NACRES:
NB.24
pore size:
0.7 μm
material:
borosilicate glass fiber (BGF) membrane (acid treated low metal)
diam.:
90 mm
sterility:
non-sterile
packaging:
pkg of 50 ea

Product Name

Whatman® acid treated low metal TCLP circles, diam. 90 mm

material

borosilicate glass fiber (BGF) membrane (acid treated low metal)

sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

Whatman 1810-090
Whatman Article No. 28418301 (US reference)

parameter

60 mL/min speed

diam.

90 mm

pore size

0.7 μm

basis weight

75 g/m2

Looking for similar products? Visit Product Comparison Guide

General description

These acid treated low metal filters are available in 47 mm, 90 mm, 110 mm, 125 mm, 142 mm, and 150 mm diameters. The 90 mm filter is required for volatile samples and use with a Zero Headspace Extractor. The 142 mm filter is typically used with non-volatile samples in an approved jar.

Toxicity Characteristic Leaching Procedure (TCLP) is an analytical test designed to determine the leaching potential in a landfill for hazardous organic and inorganic contaminants that could potentially migrate into groundwater, threatening drinking water sources.

Used for EPA Method 1311
The Whatman TCLP Filter is manufactured using a binder free borosilicate glass microfiber with a particle retention rating of 0.6 μm to 0.8 μm.

Features and Benefits

  • CLP-compliant test filters from Cytiva come in six filter sizes for accurate landfill leachate modeling and sample analysis:

Other Notes

Field of Use : For internal research use only. Products are not intended for diagnostic use or resale.

Legal Information

Whatman is a registered trademark of Cytiva

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service