920584
5-(1-(tert-Butoxycarbonyl)azetidin-3-yl)pyrazine-2-carboxylic acid
Synonym(s):
Rigid linker for PROTAC® development
About This Item
form
powder
Quality Level
reaction suitability
reagent type: linker
functional group
Boc
carboxylic acid
storage temp.
2-8°C
SMILES string
CC(C)(OC(N1CC(C2=CN=C(C(O)=O)C=N2)C1)=O)C
InChI
1S/C13H17N3O4/c1-13(2,3)20-12(19)16-6-8(7-16)9-4-15-10(5-14-9)11(17)18/h4-5,8H,6-7H2,1-3H3,(H,17,18)
InChI key
UMVYLQZBFNQMPM-UHFFFAOYSA-N
Application
Similar linkers in this series:
920487
920495
920576
ADV638390284
Other Notes
Legal Information
related product
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service