Documents
TT38440SG-EP
Eppendorf twin.tec® PCR Plates 384
skirted, PCR clean, volume 40 μL, colorless wells, green plate, pack of 25
About This Item
UNSPSC Code:
41106300
material
colorless wells
green plate
polypropylene wells
well size
384
sterility
non-sterile
feature
skirt
PCR clean
skirted
packaging
pack of 25
manufacturer/tradename
Eppendorf® 951020729 (NA)
technique(s)
PCR: suitable
volume
40 μL
working volume
40 μL
Related Categories
Features and Benefits
  • Optimized rim for barcodes and automated work processes
  • Stackable
  • Within SBS footprint recommendations
  • Ideal for most common 384-well thermocyclers, particularly for Eppendorf Mastercycler X50 and Mastercycler nexus
  • 40 µL max. well volume
  • Ideal for quantitative real-time PCR as well – OptiTrack® matrix for faster sample identification and fewer pipetting errors
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
twin.tec is a registered trademark of Eppendorf AG