Skip to Content
Merck
All Photos(1)

Key Documents

SML3480

Sigma-Aldrich

ULK-101

≥98% (HPLC)

Synonym(s):

(S)-N-(1-cyclopropyl-2,2,2-trifluoroethyl)-4-(6-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidin-3-yl)thiophene-2-carboxamide, N-[(1S)-1-Cyclopropyl-2,2,2-trifluoroethyl]-4-[6-(4-fluorophenyl)pyrazolo[1,5-a]pyrimidin-3-yl]-2-thiophenecarboxamide, ULK 101, ULK101

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16F4N4OS
CAS Number:
Molecular Weight:
460.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

SMILES string

FC(F)(F)[C@H](C1CC1)NC(C2=CC(C3=C4N(N=C3)C=C(C5=CC=C(C=C5)F)C=N4)=CS2)=O

InChI

1S/C22H16F4N4OS/c23-16-5-3-12(4-6-16)15-8-27-20-17(9-28-30(20)10-15)14-7-18(32-11-14)21(31)29-19(13-1-2-13)22(24,25)26/h3-11,13,19H,1-2H2,(H,29,31)/t19-/m0/s1

InChI key

PFZRXJIYAFANHP-IBGZPJMESA-N

Biochem/physiol Actions

Potent and selective ULK1 inhibitor that sensitizes cancer cells to nutrient stress by suppressing autophagy induction and autophagic flux.
ULK-101 is a potent and selective ULK1 inhibitor (ULK1/2 IC50 = 8.3/30 nM) that inhibits ULK1-dependent cellular Beclin 1 phosphorylation (pSer15 IC50 = 390 nM) with greater kinome selectivity (4 off-target hits using a 327-kinase panel) and potency than SBI-0206965 (IC50 = 38 nM/cell-free ULK1 kinase assay, 2.4 μM/cellular Beclion 1 pSer15). ULK-101 sensitizes cells to nutrient stress in U2OS (EC50 = 6.3 μM vs. 22.3 μM with full media) and in KRAS mutant lung cancer cultures (full media/nutrient withdrawal EC50 ratio >5) by suppressing autophagy induction and autophagic flux.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service