The flip-cover is a plus feature, like this design
HS120538
96 Well Low temp PCR rack
assorted colors, Blue, Green, Purple, Yellow, Orange, polypropylene
Synonym(s):
cold storage tube rack, freezer vial rack
About This Item
Recommended Products
material
polypropylene
feature
Holds 96 x 0.2 mL tubes (individual or strips)
packaging
pkg of 5
manufacturer/tradename
Heathrow Scientific HS120538
technique(s)
PCR: suitable
color
assorted colors, Blue, Green, Purple, Yellow, Orange
Looking for similar products? Visit Product Comparison Guide
General description
The PCR rack base is made from robust ABS construction that helps keep samples safe so you don′t have to spend extra money and time on expensive retesting. A location ID of numbers and letters allows for organized sample placement which results in easy and quick sample retrieval. The PCR rack has a closed base design that helps samples integrity by staying cold longer.
The PCR storage rack is versatile; it can be used on a bench top or for freezer storage. The compact footprint is stackable, which helps save valuable space in the lab. The PCR storage rack is offered in multiple colors for coding options.
- The lid is ambidextrous; it can be switched to accommodate left or right hand orientation
- The lid is detachable
- Holds 0.2mL tubes, individual or strip
- 96 wells 8x12 configuration
- Can be used for freezer storage down to -80°C (-112°F)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service