Skip to Content
Merck
All Photos(4)

Key Documents

HPA003328

Sigma-Aldrich

Anti-UBE2L6 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-RIG-B, Anti-Retinoic acid-induced gene B protein, Anti-UbcH8, Anti-Ubiquitin carrier protein L6, Anti-Ubiquitin- protein ligase L6, Anti-Ubiquitin/ISG15-conjugating enzyme E2 L6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IF
IHC
clone:
polyclonal
species reactivity:
human
citations:
3
technique(s):
immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

NLSSDDANVLVWHALLLPDQPPYHLKAFNLRISFPPEYPFKPPMIKFTTKIYHPNVDENGQICLPIISSENWKPCTKTCQVLEALNVLVNRPNIREPLRMDLADLLTQNPELFRKNAEEFTLR

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... UBE2L6(9246)

Immunogen

Ubiquitin/ISG15-conjugating enzyme E2 L6 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

UBE2L6 (ubiquitin-conjugating enzyme E2L 6) gene encodes a member of the E2 ubiquitin-conjugating enzyme family that catalyzes ISG15 (Ubiquitin-like protein conjugated to cellular proteins after IFN-α/β stimulation) conjugation or the attachment of ubiquitin to other proteins. Ubiquitination of proteins is an important mechanism during the degradation of abnormal or short-lived proteins. It ubiquitinates and degrades FMS-like tyrosine kinase 3 mutant and modulates leukemic cell survival.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86443

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen Zhao et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(20), 7578-7582 (2004-05-08)
Ubiquitin-(Ub) like proteins (Ubls) are conjugated to their targets by an enzymatic cascade involving an E1 activating enzyme, an E2 conjugating enzyme, and in some cases an E3 ligase. ISG15 is a Ubl that is conjugated to cellular proteins after
M Buchwald et al.
Leukemia, 24(8), 1412-1421 (2010-05-29)
The class III receptor tyrosine kinase FMS-like tyrosine kinase 3 (FLT3) regulates normal hematopoiesis and immunological functions. Nonetheless, constitutively active mutant FLT3 (FLT3-ITD) causally contributes to transformation and is associated with poor prognosis of acute myeloid leukemia (AML) patients. Histone
Tomoharu Takeuchi et al.
Journal of biochemistry, 138(6), 711-719 (2006-01-24)
ISG15 is a ubiquitin-like protein that is upregulated on treatment with interferon. ISG15 is considered to be covalently conjugated to cellular proteins through a sequential reaction similar to that of the ubiquitin conjugation system consisting of E1/E2/E3 enzymes: UBE1L and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service