Skip to Content
Merck
All Photos(1)

Documents

YL1/2

92092402, Reactivity: Anti-Tubulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

biological source

Rat x Rat Hybridoma

ImmunoDon

LOU rat spleen

Myeloma

Y3Ag1.2.3

Reactivity

Anti-Tubulin

technique(s)

cell culture | mammalian: suitable

isotype

IgG2a

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Rat Y3Ag1.2.3 x Rat LOU spleen

Cell Line Description

The antibody recognises tubulin from yeast as well as from birds and mammals. It appears to be specific for the α subunit of tubulin.

Immunogen

Yeast tubulin

Culture Medium

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service