BUA481F4-EP
Buckets for Eppendorf® A-4-81 Rotor
MTP/Flex bucket, pack of 4
All Photos(1)
About This Item
UNSPSC Code:
41103910
Recommended Products
feature
Holds x flex bucket
MTP/Flex bucket
packaging
pack of 4
manufacturer/tradename
Eppendorf® 022638840 (NA)
Eppendorf® 5810741009 (EU)
technique(s)
centrifugation: suitable
General description
Eppendorf is a well-known company in the field of life sciences. They specialize in creating and selling various tools, materials, and services used in laboratories around the world. Their product lineup includes pipettes, automated pipetting systems, dispensers, centrifuges, mixers, spectrometers, and DNA amplification tools. Eppendorf products are widely used in both academic and commercial research labs. Centrifuges and microcentrifuges find applications in various fields such as microbiology, tissue culture, molecular biology, PCR, genomics, clinical research, drug discovery, and proteomics. We provide a range of centrifuges, rotors, buckets, and adapters designed to meet your specific processing requirements. This bucket can be used for rotors A-4-81, S-4x500. It is suitable for use with IsoRack and cell culture flask adapters as well as MTP and DWP.
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)