Skip to Content
Merck
All Photos(1)

Documents

ADAP02-EP

Adapter for all Eppendorf® 1.5/2.0 mL Rotors

holds 1 x 0.2 mL PCR tubes, pack of 6 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103911

feature

holds 1 x 0.2 mL PCR tubes

packaging

pack of 6 ea

manufacturer/tradename

Eppendorf® 022636260 (NA)

parameter

16,200 × g max. RCF

technique(s)

PCR: suitable
centrifugation: suitable

compatibility

for use with Rotors 1.5/2.0 mL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Six adapters, each holding 1 PCR tube (0.2 mL, maximum diameter 6 mm). They can be used in all Eppendorf rotors holding 1.5/2.0 mL tubes.

Legal Information

Eppendorf is a registered trademark of Eppendorf AG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service