Skip to Content
Merck
All Photos(1)

Key Documents

01615

Supelco

Pullulan Standard

certified reference material, certified according to DIN, for GPC, 200,000

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41116107
E Number:
E1204
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material
for GPC

Quality Level

shelf life

limited shelf life, expiry date on the label

mol wt

Mn ~188500
Mp ~200000
Mw ~212000
Mw/Mn ~1.13

analyte chemical class(es)

oligosaccharides

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pullulan Standards are mostly used to determine molecular mass distribution of cellulosic samples. It mainly consists of polymaltotriose units linked together by α-(1→6) linkages.[1]

Application

It may be used in determination of molecular weight of hydrolysate using gel permeation chromatography, in a study performed to investigate enzymatic degradation of chitins and partially deacetlylated chitins of squid pen and shrimp shell.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Enzymatic degradation of chitins and partially deacetylated chitins.
Shigemasa, Yoshihiro, et al.
International Journal of Biological Macromolecules, 16.1, 43-49 (1994)
Chi-San Wu
Handbook Of Size Exclusion Chromatography And Related Techniques: Revised And Expanded, 336-336 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service