Skip to Content
Merck
All Photos(1)

Documents

5.31230

Sigma-Aldrich

Troponin Activator, CK-2017357

Synonym(s):

Troponin Activator, CK-2017357, 6-Ethynyl-1-(3-pentanyl)-1H-imidazo[4,5-b]pyrazin-2-ol, 6-Ethynyl-1-(pentan-3-yl)-1H-imidazo[4,5-b]pyrazin-2-ol, Fast Skeletal Troponin Activator, CK2017357, Tirasemtiv, CK-357, Tirasemtiv, CK-357, 6-Ethynyl-1-(3-pentanyl)-1H-imidazo[4,5-b]pyrazin-2-ol, 6-Ethynyl-1-(pentan-3-yl)-1H-imidazo[4,5-b]pyrazin-2-ol, Fast Skeletal Troponin Activator, CK2017357

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N4O
CAS Number:
Molecular Weight:
230.27
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

beige

solubility

DMSO: 25 mg/mL

storage temp.

2-8°C

InChI

1S/C12H14N4O/c1-4-8-7-13-10-11(14-8)16(12(17)15-10)9(5-2)6-3/h1,7,9H,5-6H2,2-3H3,(H,13,15,17)

InChI key

RSQGZEAXODVTOL-UHFFFAOYSA-N

General description

An orally bioavailable, non-toxic imidazopyrazinol compound that directly and selectively binds to the troponin complex in fast skeletal muscle (Kd = 40 nM) and sensitizes it to calcium. Its binding site is believed to be between two or more troponin subunits. Exhibits very modest affinity for slow skeletal muscle troponin (Kd = 3.8 µM) and has no measurable affinity for cardiac muscle troponin. Effectively stimulates myosin ATPase activity in fast skeletal muscle (EC50 = 390 nM), but has significantly reduced effect on mixed fiber type (EC50 = 770 nM in bovine rectus abdominis muscle). Slows the first order rate constant for Ca2+ release by about 70% (from 14.7 s-1 to 4.0 s-1) and shifts the plot of myosin ATPase relationship to calcium leftwards in fast skeletal muscles without increasing the maximum recorded force or the shape of the curve. Shown to improve muscle function in a rodent model of neuromuscular disease where neural input is diminished.
An orally bioavailable, non-toxic imidazopyrazinol compound that directly and selectively binds to the troponin complex in fast skeletal muscle (Kd = 40 nM) and sensitizes it to calcium. Its binding site is believed to be between two or more troponin subunits. Exhibits very modest affinity for slow skeletal muscle troponin (Kd = 3.8 µM) and has no measurable affinity for cardiac muscle troponin. Effectively stimulates myosin ATPase activity in fast skeletal muscle (EC50 = 390 nM), but has significantly reduced effect on mixed fiber type (EC50 = 770 nM in bovine rectus abdominis muscle). Slows the first order rate constant for Ca2+ release by about 70% (from 14.7 s-1 to 4.0 s-1) and shifts the plot of myosin ATPase relationship to calcium leftwards in fast skeletal muscles without increasing the maximum recorded force or the shape of the curve. Shown to improve muscle function in a rodent model of neuromuscular disease where neural input is diminished.

Please note that the molecular weight for this compound is batch-specific due to variable water content.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
troponin complex
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C).

Other Notes

Gordon, P., et al. 2013. Expert Opin. Phamacother.14, 1907.
Miciak, J.J., et al. 2013. Muscle Nerve.48, 279.
Russell, A.J., et al. 2012. Nat. Med.18, 452.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service