This product is distributed on behalf of Büchi as 1 each.
Z402923
Vacuum seals for Büchi® Rotavapor® evaporators
KD 22 stationary, for R114/R124/R100
Synonym(s):
Büchi Rotavapor Seals, Seals for Büchi Evaporators, Vacuum Seals for Rotavapor
About This Item
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Büchi 000636
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Compatible with BUCHI R-100 and R-3 Rotavapors.
- OEM seals ensure proper fit and performance.
- Offer high chemical resistance.
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
how many seals are included in the product
1 answer-
Helpful?
-
-
Is product Z402923 compatible with R111?
1 answer-
Yes, Z402923, Vacuum seal is compatible with Buchi R-111 rotary evaporator.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service