928801
Barrel and piston
3 mL, amber, suitable for 3D bioprinters, 100/box
Synonym(s):
3D Bioprinting consumables, Syringe barrels
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
42142600
NACRES:
NA.25
Recommended Products
description
3mL (3 cc) barrels (amber) and pistons (white) (50/box),
Barrel Diameter 11.1 mm (0.44′′), Length 73.0 mm (2.88′′)
Looking for similar products? Visit Product Comparison Guide
Application
Empty cartridges for 3D Bioprinting application. The precision fit between barrel and piston ensures consistent fluid deposit. Wiper piston improves fluid control and keeps fluid from dripping.
End and tip caps are sold separately.
End and tip caps are sold separately.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service