Skip to Content
Merck
All Photos(1)

Key Documents

928593

Sigma-Aldrich

Optical Readout Reaction Chamber Chip

Fluidic 1068, COP-black

Synonym(s):

Cyclic olefin polymer, Microfluidic chip

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
42142600
NACRES:
NA.25

description

Microfludic chip x1

Looking for similar products? Visit Product Comparison Guide

Application

The Optical Readout Reaction Chamber Chip, Fluidic 1068 COP-black microfluidic chip has eight chambers suitable for optical readouts. The chip is designed with reaction chambers on the top side of the chip, making it especially suited for readouts done using an upright microscope.

Features and Benefits

  • Reaction chambers suitable for colorimetric, luminescent or fluorescent optical readouts
  • Designed to be used with an upright microscope


Chip Properties:
  • Mini Luer Interface
  • Material: Cyclic olefin polymer (COP)
  • Material color: Black
  • 8 reaction chambers
  • Chamber volume: 20 µl

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service