Skip to Content
Merck
All Photos(1)

Documents

526428

Sigma-Aldrich

2-Chloro-3,6-difluorobenzaldehyde

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H2(F)2CHO
CAS Number:
Molecular Weight:
176.55
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

46-50 °C (lit.)

SMILES string

Fc1ccc(F)c(C=O)c1Cl

InChI

1S/C7H3ClF2O/c8-7-4(3-11)5(9)1-2-6(7)10/h1-3H

InChI key

KPYBGKSDBNMHTO-UHFFFAOYSA-N

General description

2-Chloro-3,6-difluorobenzaldehyde is a trisubstituted benzaldehyde. Fourier-transform, laser Raman (in the range of 3500-50cm-1) and infrared (in the range of 4000-400cm-1) spectral analyses of 2-chloro-3,6-difluorobenzaldehyde have been reported.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vibrational assignments and effect of aldehyde rotation on substituents in some trisubstituted benzaldehydes.
Hiremath CS and Tonannavar J.
Spectrochimica Acta. Part A, Molecular and Biomolecular Spectroscopy, 73(2), 388-397 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service