Skip to Content
Merck
All Photos(1)

Key Documents

488976

Sigma-Aldrich

(±)-Cotinine-(methyl-d3)

99 atom % D

Synonym(s):

1-(Methyl-d3)-5-(3-pyridinyl)-2-pyrrolidinone, Deuterated (±)-cotinine solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10D3H9N2O
CAS Number:
Molecular Weight:
179.23
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

99 atom % D

Quality Level

Assay

99% (CP)

form

solid
(viscous liquid or solid)

technique(s)

mass spectrometry (MS): suitable

color

colorless to yellow

bp

250 °C/150 mmHg (lit.)

mp

40-42 °C (lit.)

mass shift

M+3

storage temp.

2-8°C

SMILES string

[2H]C([2H])([2H])N1C(CCC1=O)c2cccnc2

InChI

1S/C10H12N2O/c1-12-9(4-5-10(12)13)8-3-2-6-11-7-8/h2-3,6-7,9H,4-5H2,1H3/i1D3

InChI key

UIKROCXWUNQSPJ-FIBGUPNXSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service