21738
Valco® Precision Sampling syringe, Magnum series
volume 500 mL, removable needle, needle L × O.D. × I.D. 2 in. × 0.035 in. × 0.023 in.
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
bevel tip needle
glass syringe
description
Magnum Series
feature
removable needle
packaging
pkg of 1 ea
manufacturer/tradename
VICI 060042
needle L × O.D. × I.D.
2 in. × 0.035 in. × 0.023 in.
volume
500 mL
Related Categories
General description
Magnum Series - Removable Needle: This Pressure-Lok series syringe allows sampling and storage of samples up to 500 mL. A low-dead volume, removable needle is used, and a rotary valve allows on-site sampling for later analysis. It is ideal for collection, trace analysis and sample concentration of environmental contaminants.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)