Key Documents
Z760994
PCV cell counting tubes
no cap or graduations, for easy read ruler
Synonym(s):
Cell Counting Tubes, PCV Counting Tubes, Tubes for Cell Counting
About This Item
UNSPSC Code:
41121706
NACRES:
NB.22
material
(PCV)
colorless tube
conical bottom
straight neck
sterility
non-sterile
packaging
pack of 50 ea
manufacturer/tradename
TPP 87007
availability
not available in (available only in North America)
General description
The packed cell volume (PCV) tube is a quick and reproducible measuring technique of the cell volume that is suitable to monitor cell growth (cell mass).
Features and Benefits
  • Present with or without scale.
  • Fits in microcentrifuges.
  • Caps available if required.
  • Ideal to monitor cell growth
recommended
Product No.
Description
Pricing
Choose from one of the most recent versions:
Certificates of Analysis (COA)