TMO6370-0004
Biohazard waste container
volume 5.5 L, polypropylene, pack of 1 ea
Synonym(s):
autoclave waste disposal, biohaz waste bag, biohazardous disposal bags, waste bags
All Photos(1)
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Recommended Products
material
polypropylene
packaging
pack of 1 ea
manufacturer/tradename
Nalge-Nunc 6370-0004
volume
5.5 L
5500 mL
Related Categories
General description
Biohazardous Waste Containers are ideal autoclavable polypropylene containers for secondary containment of biohazardous waste materials.
Convenient one-step sterilization; no need to remove bag. Port opening in cover allows one-handed disposal of materials; limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust, dent or puncture.
Convenient one-step sterilization; no need to remove bag. Port opening in cover allows one-handed disposal of materials; limits exposure to infectious waste. Holds standard sized autoclave bags. Will not rust, dent or puncture.
Choose from one of the most recent versions:
Certificates of Analysis (COA)