TMO374110
Nunc® Coded Cryobank Vials in black low rack
polypropylene, capacity 1.0 mL, self-standing natural, sterile, green cap, case of 960 ea
Synonym(s):
coded cryobank vial storage, cryostorage
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
green cap
polypropylene
self-standing natural
sterility
sterile
packaging
case of 960 ea
manufacturer/tradename
Nunc 374110
technique(s)
cryopreservation: suitable
capacity
1.0 mL
Looking for similar products? Visit Product Comparison Guide
General description
Related applications: Cell Culture Plastics
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service