EP5702710008
Eppendorf® Adapter for 90 mL rectangular bucket for use with standard tubes
Holds 5 - 7 mL tubes, pack of 2 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
feature
Holds 5 - 7 mL tubes
packaging
pack of 2 ea
manufacturer/tradename
Eppendorf® 5702710008
technique(s)
centrifugation: suitable
compatibility
for use with 90 mL rectangular buckets
Looking for similar products? Visit Product Comparison Guide
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service