CLS6796600D
Corning® hotplate with digital display
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 10 in. × 10 in., CE compliant
Synonym(s):
stirrers
All Photos(2)
About This Item
Recommended Products
feature
CE compliant
packaging
case of 1
manufacturer/tradename
Corning 6796-600D
availability
not available in USA and Canada
parameter
5-550 °C temp. range
plate L × W
10 in. × 10 in.
General description
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards
- 2 year warranty
Legal Information
Corning is a registered trademark of Corning, Inc.