Key Documents
CLS6634
Corning® Step-R Repeating Pipettor Syringe Tips
volume (25.0 mL), sterile
Synonym(s):
pipet tips
About This Item
UNSPSC Code:
41121600
NACRES:
NB.02
sterility
sterile
packaging
pack of 25 ea
manufacturer/tradename
Corning 6634
volume
(25.0 mL)
Related Categories
General description
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Legal Information
Corning is a registered trademark of Corning, Inc.