CLS354245
Corning® Collagen IV
from human, pack of 0.25 mg
Synonym(s):
extracellular matrix
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
human
packaging
pack of 0.25 mg
manufacturer/tradename
Corning 354245
Looking for similar products? Visit Product Comparison Guide
General description
Corning Collagen IV, human, 0.25mg, is used as a thin coating on tissue-culture surfaces to promote cell attachment and proliferation and to study the collagen′s effects on cell behavior.
Source
Human placenta. NOTE: Source material tested for HBsAG, for anti-HIV-1 and anti-HIV-2, and anti-HCV virus-1 and virus-2.
Quality
Electrophoretically homogeneous (SDS-PAGE)
Tested and found negative for bacteria, fungi, and mycoplasma
Preparation and Storage
Stable for at least three months at -70°C. Avoid multiple freeze-thaws.
Handling
Recommended concentration - 1-5 μg/cm² of growth surface depending on cell type
Formulation
Frozen in 10 mM Acetic Aci
Source
Human placenta. NOTE: Source material tested for HBsAG, for anti-HIV-1 and anti-HIV-2, and anti-HCV virus-1 and virus-2.
Quality
Electrophoretically homogeneous (SDS-PAGE)
Tested and found negative for bacteria, fungi, and mycoplasma
Preparation and Storage
Stable for at least three months at -70°C. Avoid multiple freeze-thaws.
Handling
Recommended concentration - 1-5 μg/cm² of growth surface depending on cell type
Formulation
Frozen in 10 mM Acetic Aci
Legal Information
Corning is a registered trademark of Corning, Inc.
Storage Class Code
12 - Non Combustible Liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service