Skip to Content
Merck
All Photos(2)

Key Documents

CLS352037

Corning® Falcon® Round Bottom Test Tubes

capacity 16 mL, cap, screw, sterile, individually wrapped, case of 500

Synonym(s):

culture tube

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121700
NACRES:
NB.13

material

round polystyrene

sterility

sterile

feature

cap screw
individually wrapped

packaging

case of 500

manufacturer/tradename

Corning 352037

capacity

16 mL

Looking for similar products? Visit Product Comparison Guide

Application

Liquid Handling

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service