Skip to Content
Merck
All Photos(1)

Key Documents

Y1

85062807, mouse adrenal gland (cortex), Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

Product Name

Y1, 85062807

biological source

mouse adrenal gland (cortex)

growth mode

Adherent

karyotype

2n = 40

morphology

Epithelial

products

Steroid hormone

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Mouse LAF1 adrenal cortex tumour

Cell Line Description

This cell line is a cloned strain from an adrenal cortex tumour cell line of a male LAF1 mouse. This strain was isolated after alternate passage in tumours in vivo and in cultures in vitro . The isolated clones are reported by the original depositor to produce steroid hormones at a high rate (4ug/mg/hr) under ACTH stimulation and cells round up within minutes of ACTH treatment. The hormone produced is reported to be ketosteroid with a hydroxy group at the carbon 20 rather than the usual ketosteroid configuration. However, a laboratory independent to the depositor has since (Dec 2000) tested this steroid producing property using the same method and after 1 hour of stimulation could report no rounding up or change in morphology and a very low cAMP measurement. This laboratory did find that the cells round up when exposed to forskolin (acting through adenylate cyclase) and the original depositor reported morphological changes in response to cholera toxin.

Application

Cloning and hormone studies

Culture Medium

Ham′s F10 + 2mM Glutamine + 15% Horse Serum (HS) + 2.5% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:3 to 1:6 i.e. seeding at 2-4x10,000 cells/cm2 using 0.25% trypsin/EDTA; 5% CO2; 37°C.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service