AXYTE204Y
Pipette tips for Eppendorf® pipettors
size 200 μL, yellow, pkg of 10x1000tips/cs
Synonym(s):
pipet tips
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Axygen Inc TE-204-Y
Corning
packaging
pkg of 10x1000tips/cs
size
200 μL
color
yellow
Looking for similar products? Visit Product Comparison Guide
Other Notes
Browse all pipette tips
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service