AXYMCT200LC
MAXYMum Recovery® microtubes
size 2.0 mL, pkg of 10x250tubes/cs
Synonym(s):
centrifuge tubes
About This Item
Recommended Products
material
cap (snap cap)
conical bottom tube
polypropylene
polypropylene cap
sterility
non-sterile
feature
RNase and DNase free
endotoxin free
graduations
manufacturer/tradename
Axygen Inc MCT-200-L-C
Corning
packaging
pkg of 10x250tubes/cs
size
2.0 mL
color
clear
fitting
cap (push fit)
suitability
suitable for (centrifugation)
binding type
(ultra-low retention)
Looking for similar products? Visit Product Comparison Guide
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service