Skip to Content
Merck
All Photos(1)

Key Documents

AXYHS400

Axygen PlateMax heat sealing film for compound storage

peelable, 100 films/unit, 5 units/case

Synonym(s):

sealing mat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.13

sterility

non-sterile

form

film

manufacturer/tradename

Axygen Inc HS-400
Corning

General description

  • Axygen® sealing films are used in PCR and Real-time PCR to ELISA and cell culture applications.
  • PlateMax® heat sealing films offer high-temperature resistance.
  • Made from polyester/aluminum
  • Aluminum material is for light-sensitive samples and polyester-based sealing films are ideal for short-term storage/incubation.
  • Compatible with the Axygen® PlateMax® Semi-Automated Plate Sealer (HS-1120, HS-1230).

Legal Information

Axygen is a registered trademark of Corning, Inc.
PlateMax is a registered trademark of Axygen, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service