Skip to Content
Merck
All Photos(1)

Key Documents

E2451000

Etilefrine hydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-Ethylamino-1-(3-hydroxyphenyl)-1-ethanol hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO2 · HCl
CAS Number:
Molecular Weight:
217.69
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

etilefrine

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C10H15NO2.ClH/c1-2-11-7-10(13)8-4-3-5-9(12)6-8;/h3-6,10-13H,2,7H2,1H3;1H

InChI key

KTNROWWHOBZQGK-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Etilefrine hydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Management of thoracic duct injuries after oesophagectomy (Br J Surg 2001: 88: 1442-8). Letter 1.
P Guillem et al.
The British journal of surgery, 89(6), 809-809 (2002-05-25)
Ioannis E Platis et al.
Thoracic surgery clinics, 16(3), 209-214 (2006-09-29)
Chylothorax is a rare complication of pulmonary resection. It requires prompt treatment, which is initially conservative. This treatment consists of drainage, nutritional support, and measures to diminish chyle flow. Surgical intervention is indicated when conservative management is ineffective. Delay in
Iheanyi Okpala
Expert opinion on investigational drugs, 15(8), 833-842 (2006-07-25)
Developments in the treatment of sickle cell disease (SCD) have not kept pace with advances in understanding the pathophysiology of this haemoglobinopathy. Drugs undergoing preclinical and clinical assessment for the therapy of these globin gene disorders are discussed in this
C Schwerk et al.
Klinische Padiatrie, 221(2), 93-96 (2009-02-10)
We report here on a case of etilefrinhydrochloride tablet intoxication in suicidal intention in a female adolescent. Tablet ingestion resulted in the formation of a tablet conglomerate in the stomach, which could neither be effectively treated with activated charcoal nor
Michele Brignole
Journal of cardiovascular electrophysiology, 14(9 Suppl), S64-S69 (2003-09-03)
Evidence for therapy of neurally mediated syncope is generally weak. Many drugs have been used for the treatment of vasovagal syncope (beta-blockers, disopyramide, scopolamine, clonidine, theophylline, fludrocortisone, ephedrine, dihydroergotamine, etilefrine, midodrine, clonidine, serotonin reuptake inhibitors, enalapril). In general, although the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service