Skip to Content
Merck
All Photos(2)

Key Documents

33448

Supelco

Nequinate

VETRANAL®, analytical standard

Synonym(s):

Methyl 7-(benzyloxy)-6-butyl-1,4-dihydro-4-oxo-3-quinolinecarboxylate, Methyl benzoquate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H23NO4
CAS Number:
Molecular Weight:
365.42
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCCCc1cc2C(=O)C(=CNc2cc1OCc3ccccc3)C(=O)OC

InChI

1S/C22H23NO4/c1-3-4-10-16-11-17-19(23-13-18(21(17)24)22(25)26-2)12-20(16)27-14-15-8-6-5-7-9-15/h5-9,11-13H,3-4,10,14H2,1-2H3,(H,23,24)

InChI key

NNOPDLNHPOLRRE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J E Peeters et al.
Research in veterinary science, 35(2), 211-216 (1983-09-01)
For 15 months the anticoccidial effect of 200 ppm clopidol/methyl benzoquate and of 50 ppm robenidine, and the development of immunity against five different species of Eimeria were followed in a closed rabbit population. In unmedicated rabbits, oocyst output decreased
Influence of the anticoccidials clopidol/methylbenzoquate and robenidine on fertility and progeny of female rabbits.
H Varewyck et al.
The British veterinary journal, 140(2), 202-206 (1984-03-01)
Activity of clopidol/methylbenzoquate, robenidine and salinomycin against hepatic coccidiosis in rabbits.
J E Peeters et al.
Zentralblatt fur Veterinarmedizin. Reihe B. Journal of veterinary medicine. Series B, 29(3), 207-218 (1982-04-01)
L P Joyner et al.
Research in veterinary science, 34(1), 64-67 (1983-01-01)
The activity of sulphaquinoxaline, robenidine, methyl benzoquate, clopidol and a mixture of methyl benzoquate and clopidol (Lerbek; Dow), was studied in rabbits infected with hepatic coccidiosis due to Eimeria stiedai. Growth inhibition, oocyst production and the activity in the serum
Determination of methyl benzoquate in poultry feedingstuffs using high-performance liquid chromatography.
G H Merson et al.
The Analyst, 110(7), 761-764 (1985-07-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service