Skip to Content
Merck
All Photos(2)

Key Documents

MABC1031

Sigma-Aldrich

Anti-TNFSF13 (APRIL) Antibody, clone A3D8

clone A3D8, from hamster(Armenian)

Synonym(s):

A proliferation-inducing ligand, APRIL, CD256, TALL-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

biological source

hamster (Armenian)

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A3D8, monoclonal

species reactivity

human, mouse

technique(s)

flow cytometry: suitable

isotype

IgG

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

mouse ... Tnfsf13(69583)

Related Categories

General description

Tumor necrosis factor ligand superfamily member 13 (UniProt: Q9D777; also known as A proliferation-inducing ligand; APRIL; CD256) is encoded by the Tnfsf13 (also known as APRIL) gene (Gene ID: 69583) in mouse. APRIL is a secreted cytokine of the tumor necrosis family and is present in T cells, B cells, dendritic cells, monocytes, and macrophages. April exists as a functional homotrimer, but can form functional heterotrimers with B cell activating factor (BAFF). APRIL stimulates T and B cell proliferation and it exerts T and B cell regulatory effects by binding to TNFRSF17 (BCMA) and TNFRSF13B (TACI) receptors. It is also involved in monocyte/macrophage-mediated immunological processes. It also plays a role in the regulation of tumor cell growth and can induce apoptosis through its interaction with other TNF receptor family proteins such as TNFRSF6/FAS and TNFRSF14/HVEM. APRIL can serve as as a potential biomarker and its serum levels are shown to be elevated for certain cancers and expression levels elevated in cancer tissues.

Specificity

Clone A3D8 detects TNFSF13/APRIL in human peripheral blood mononuclear cells (PBMC).

Immunogen

A full length recombinant fusion protein of murine maltose binding protein (MBP) and soluble APRIL.

Application

Detect TNFSF13 using this armenian hamster monoclonal Anti-TNFSF13 (APRIL) antibody, clone A3D8, Cat. No. MABC1031, validated for use in Flow Cytometry.
Flow Cytometry Analysis: 1 µg of this antibody from a representative lot detected TNFSF13 in one million RAW264.7 cells.
Research Category
Apoptosis & Cancer

Quality

Evaluated by Flow Cytometry in human PBMC cells.

Flow Cytometry Analysis: 1 µg of this antibody detected TNFSF13 in human PBMC cells.

Target description

26.89 kDa calculated.

Physical form

Format: Purified
Protein G purified
Purified armenian hamster monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service