IPAKGARH2
IPAK Gard® Pretreatment Pack
This product is replaced with IPAKGARDH2. Search to Explore!
Synonym(s):
IPAK gard® H 10-15 Pretreatment pack
About This Item
Quality Level
feature
For optimal protection of the reverse osmosis membrane
packaging
pkg of 1 unit
manufacturer/tradename
IPAK Gard®
H
39.5 cm (15.5 in.)
input
feed water nature potable tap water (with hardness (as CaCO3) > 300 ppm)
compatibility
for use with Milli-Q® IQ 7010
for use with Milli-Q® IQ 7015
Looking for similar products? Visit Product Comparison Guide
General description
For use with Milli-Q® IQ 7010, Milli-Q® IQ 7015, Milli-Q® IX 7010 and Milli-Q® IX 7015 water purification systems.
Application
Features and Benefits
- Activated carbon protects against free chlorine.
- Polyphosphate beads protect against scaling and precipitates.
- Depth filter protects against particulates and colloids.
- 63% of the parts suppliers are ISO 14001 environmental management system-certified.
Legal Information
also commonly purchased with this product
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service