Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
purified antibody
serum
antibody product type
primary antibodies
clone
polyclonal
mol wt
calculated mol wt 67.57 and 48.74 kDa
observed mol wt ~62 and 100 kDa
purified by
(Unpurified)
species reactivity
human
species reactivity (predicted by homology)
rat, mouse
packaging
antibody small pack of 100 μL
technique(s)
ChIP: suitable
western blot: suitable
isotype
IgG
epitope sequence
C-terminal
Protein ID accession no.
UniProt accession no.
shipped in
dry ice
target post-translational modification
unmodified
Gene Information
human ... runx1t1> RUNX1T1(862)
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in Kasumi-1; t(8;21) cells expressing AML1-ETO.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected AML1-ETO in Kasumi-1 ; t(8;21) cells expressing AML1-ETO.
Tested Applications
Immunoprecipitation Analysis: A representative lot immunoprecipitated AML1-ETO in Kasumi-1; t(8;21) cells expressing AML1-ETO (Courtesy of Dr Jinsong Zhang @ Saint Louis University).
Chromatin immunoprecipitation (ChIP): A representative lot detected AML1-ETO in Kasumi-1; t(8;21) cells expressing AML1-ETO (Courtesy of Dr Jinsong Zhang @ Saint Louis University).
ChIP-seq: A representative lot detected AML1-ETO in ChIP-seq applications (Guo, Chun., et. al. (2020). J Biol Chem. 295(13); 4212-4223.
Chromatin Immunoprecipitation (ChIP) Analysis: A representative lot detected AML1-ETO in Chromatin Immunoprecipitation applications (Guo, Chun., et. al. (2020). J Biol Chem. 295(13):4212-4223; Steinauer, Nickolas., et. al. (2019). Blood Adv. 3(9): 1379-1393).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service