Skip to Content
Merck
All Photos(1)

Key Documents

444284

Sigma-Aldrich

MMP-2/MMP-3 Inhibitor III, PD166793

The MMP-2/MMP-3 Inhibitor III, PD166793, also referenced under CAS 199850-67-4, controls the biological activity of MMP-2/MMP-3. This small molecule/inhibitor is primarily used for Protease Inhibitors applications.

Synonym(s):

MMP-2/MMP-3 Inhibitor III, PD166793, (S)-2-(4ʹ-Bromo-biphenyl-4-sulfonylamino-3-methylbutyric acid)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H18BrNO4S
CAS Number:
Molecular Weight:
412.30
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 40 mg/mL
ethanol: 40 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[S](=O)(=O)(N[C@@H](C(C)C)C(=O)O)c1ccc(cc1)c2ccc(cc2)Br

InChI

1S/C17H18BrNO4S/c1-11(2)16(17(20)21)19-24(22,23)15-9-5-13(6-10-15)12-3-7-14(18)8-4-12/h3-11,16,19H,1-2H3,(H,20,21)/t16-/m0/s1

InChI key

GJOCABIDMCKCEG-INIZCTEOSA-N

General description

A cell-permeable biphenylsulfonylvaline compound that acts as a potent inhibitor against MMP-2, -3, and -13 (IC50 = 47, 12, and 8 nM, respectively) and a weaker inhibitor against AMP deaminase (20% inhibition at 0.1 µM), MMP-1, -7, -9, and -14 (IC50 = 6.1, 7.2, 7.9, and 0.24 µM, respectively). Shown to offer therapeutic benefits in vivo in various animal models of heart failure and diabetes.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
MMP-2, -3, and -13
Product does not compete with ATP.
Reversible: no
Target IC50: 47, 12, and 8 nM, against MMP-2, -3, and -13, respectively

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Paolocci, N., et al. 2006. J. Pharm. Exp. Ther.317, 506.
Zhou, Y.P., et al. 2005. Diabetes54, 2612.
Peterson, J.T., et al. 2001. Circulation103, 2303.
Spinale, F.G., et al. 1999. Circ. Res.85, 364.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Select different protease inhibitor types based on your needs to prevent protein degradation during isolation and characterization and safeguard proteins in sample prep.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service