Skip to Content
Merck
All Photos(1)

Key Documents

401954

Sigma-Aldrich

Goat Anti-Human IgG, H & L Chain Specific

Calbiochem®, from goat

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.77

biological source

goat

Quality Level

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

contains

≤0.1% sodium azide as preservative

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

ambient

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Immunoaffinity purified goat polyclonal antibody. Recognizes human IgG, heavy and light chains.
This Goat Anti-Human IgG, H & L Chain Specific is validated for use in Immunoelectrophoresis for the detection of Human IgG, H & L Chain Specific.

Application

Immunoelectrophoresis (see comments)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 10 mM PBS, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Monospecific for human IgG, heavy and light chains as determined by immunoelectrophoresis against normal human serum. Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service