Skip to Content
Merck
All Photos(1)

Key Documents

890899P

Avanti

DODMA

Avanti Research - A Croda Brand

Synonym(s):

1,2-dioleyloxy-3-dimethylaminopropane; N,N-Dimethyl-2,3-bis[(9Z)-9-octadecen-1-yloxy]-1-propanamine, Avanti Polar Lipids

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C41H81NO2
Molecular Weight:
620.09
UNSPSC Code:
12352211
NACRES:
NA.79

biological source

synthetic

Assay

>99 (TLC)

form

powder

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

cationic lipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

O(CCCCCCCCC=CCCCCCCCC)CC(OCCCCCCCCC=CCCCCCCCC)CN(C)C

General description

Cationic lipids like DODMA are recognized as non-viral transporters for gene therapy. DODMA is employed in the creation of cationic liposomes or lipid nanoparticles (LNP) that require a neutral or minimal zeta potential. DODMA exhibits high efficacy in encapsulating nucleic acids because it can temporarily lower the pH.

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service