Skip to Content
Merck
All Photos(1)

Key Documents

Z516090

Sigma-Aldrich

Schlenk reaction and storage tube

male joint: ST/NS 14/20, capacity 50 mL

Synonym(s):

storage tube, storage tubes, tube flask, tube storage

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

material

colorless cap
colorless tube
round bottom

Joint

male joint: ST/NS14/20

feature

graduations: no

capacity

50 mL

Looking for similar products? Visit Product Comparison Guide

General description

2 mm glass stopcock. The side-arm accommodates a septum (use Z565695 or Z565679). Designed for small-scale manipulation of air and moisture-sensitive reagents. Thermoplastic screw caps with Viton O-rings usable to 140°C.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service