Key Documents
Z218693
Clamps
fits flask, 1,000 mL
Synonym(s):
flask clamp, flask lab, lab flask, reaction flasks
About This Item
UNSPSC Code:
41122413
NACRES:
NB.43
feature
fits flask 1,000 mL
manufacturer/tradename
Ace Glass 650806
General description
Two-piece, black anodized aluminum clamp has three wing nuts to secure Ace reaction flask to head. Clamps also fit Pyrex reaction kettles.
Choose from one of the most recent versions:
Certificates of Analysis (COA)