Key Documents
Z187224
Sigma-Aldrich
Lecture-bottle control valve with compression fitting
tube outlet O.D. 1/4 in., stainless steel
Synonym(s):
Lecture Bottle Control Valve
About This Item
UNSPSC Code:
41121800
NACRES:
NB.51
material
stainless steel
tube outlet O.D.
1/4 in.
General description
CGA 110 requires CGA 110 PTFE sealing washer.
Warning
WARNING: Gas-control valves do not control pressure in a closed system. Regulator or relief valves should be used to prevent the buildup of unsafe pressures that could lead to explosion.
Choose from one of the most recent versions:
Certificates of Analysis (COA)