Key Documents
Z173096
Sigma-Aldrich
Solvent storage flask with glass stopcock on side-arm
capacity 250 mL, female joint: ST/NS 14/20
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
material
borosilicate glass
round bottom
two-neck
label
S
Joint
female joint: ST/NS14/20
sterility
non-sterile
feature
closure type female Joint: ST/NS
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
capacity
250 mL
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Articles
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.