Z120677
Aldrich® wide-mouth dewar flasks
plastic mesh housing, capacity 1,900 mL
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
borosilicate glass
wide-mouth
Joint
Joints neck (120)
sterility
non-sterile
packaging
pack of 1
manufacturer/tradename
Sigma-Aldrich
I.D.
12.0 cm
capacity
1,900 mL
depth × Overall H
19.0 cm × 25.2 cm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Aldrich® wide-mouth dewar flasks contain borosilicate glass with protective aluminum or plastic mesh housing and extended aluminum base weighted for stability.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service