Key Documents
Z115053
Sigma-Aldrich
Vials, screw-top V-Vials® with solid top cap
volume 0.3 mL, thread for 13-425, diam. × H 13 mm × 35 mm, pkg of 12 ea
About This Item
UNSPSC Code:
41122001
NACRES:
NB.11
material
PTFE-faced styrene-butadiene rubber liner
black phenolic solid cap
clear glass vial
feature
closure type black phenolic solid top cap
closure type screw top vial
packaging
pkg of 12 ea
diam. × H
13 mm × 35 mm
volume
0.3 mL
fitting
thread for 13-425
General description
  • Graduated.
  • Autoclavable borosilicate USP Type 1 glass.
  • PTFE-faced, styrene-butadiene rubber liner.
Legal Information
V-Vial is a registered trademark of Wheaton Industries, Inc.
related product
replacement part
Choose from one of the most recent versions:
Certificates of Analysis (COA)