SYNF128000
Synthware™ single-neck flat-bottom flask
capacity 1,000 mL, joint: ST/NS 29/42
Synonym(s):
Synthware F128000, glass round flask
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.51
Recommended Products
material
borosilicate glass flask
flat bottom
single-neck
Joint
joint: ST/NS29/42
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F128000
capacity
1,000 mL
Related Categories
General description
Heavy wall single neck, flat bottom flask with an outer joint. Heavy wall design. All sizes listed below are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)