Skip to Content
Merck
All Photos(1)

Key Documents

SLW718342R

Pyrex® Pestle

glazed finish, L 110 mm

Synonym(s):

Pyrex, SciLabware 718342R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.35

Pricing and availability is not currently available.

material

borosilicate glass

manufacturer/tradename

718342R
SciLabware

L

110 mm

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA020610HPA036414SAB1410503
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

-

UniProt accession no.

Q4LDE5

UniProt accession no.

Q4LDE5

UniProt accession no.

Q8TER0

UniProt accession no.

Q9ULZ2

General description

  • Ideal for crushing solids into powders for easier handling
  • Manufactured from Pyrex borosilicate glass for highest resistance to thermal shock and chemical corrosion
  • Available in a rough or glazed finish
  • For use with Pyrex Mortars

Legal Information

PYREX is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service