SLW3722/06M
Pyrex® Büchner filter funnels with sintered disc
4000 mL, disc diam. 120 mm, porosity grade 3
Synonym(s):
Pyrex, SciLabware 3722/06M
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
manufacturer/tradename
3722/06M
SciLabware
capacity
4000 mL
4000 mL
disc diam.
120 mm
pore size
grade 3 porosity
Looking for similar products? Visit Product Comparison Guide
General description
The 175mm sintered discs fitted to these funnels are manufactured from the same 3.3 expansion borosilicate glass as the funnel bodies, making these funnels suitable to be used with a wide range aggressive chemicals.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service